missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ HDPE Serum Bottles
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
120 mL
Description
- Manufactured from high density polyethylene
- Widely used for animal health products and storage of non-medical products
- Accepts standard rubber stoppers and aluminum seals

Specifications
Specifications
Capacity | 0.5 oz., 15 mL |
Capacity (Metric) | 15 mL |
Capacity (English) | 0.5 oz. |
Color | Natural |
Diameter (English) | 1.1 in. |
Diameter (English) Inner Neck | 0.78 in. |
Diameter (Metric) Outer Neck | 20 mm |
Diameter (Metric) Inner Neck | 13 mm |
Material | HDPE |
Height (Metric) | 56 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ HDPE Serum Bottles >
Spot an opportunity for improvement?Share a Content Correction