missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Headspace Seals with Pressure Release

Brand:  DWK Life Sciences W224215

Additional Details : Weight : 0.27220kg

Product Code. 12309919

  • £164.00 / Pack of 100
Estimated Shipment: 13-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

20 mm
Designed specifically for Glass 20mm headspace autosampler vials
Seal, Headspace
Natural Gray/Silver
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers