missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ HPLC Mobile-Phase Reservoirs
Suitable for use as mobile phase and filtration reservoirs, as well as general sample and buffer storage
Brand: DWK Life Sciences 264711
This item is not returnable.
View return policy
Description
- Manufactured from Wheaton 33™ low extractable borosilicate glass, which meets ASTM Type I, Class A and USP Type I standards
- Plastic safety coating with an integral UV-blocking agent helps retain glass and liquid in case of breakage and block virtually all UV light up to 385nm
- Screw thread: GL 45
- Autoclavable up to 121°C at 15-20 psig for 15-20 minutes
- Flat bottom
Specifications
USP Type I and ASTM E 438 Type I, Class A | |
5000 mL | |
HPLC Mobile-phase Reservoir | |
12.51 in. | |
186 mm |
7.32 in. | |
Glass | |
Ideal for mobile phase or filtration reservoir | |
318 mm | |
Flat |