Learn More
DWK Life Sciences Wheaton™ Kuderna-Danish Evaporative Concentrator With Clear-Seal™ Joints
For concentration of trace amounts of analyte in organic solvent
Brand:  DWK Life Sciences 297883
Product Code. 12498932
  • £391.00 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear seal joint
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kuderna-Danish Evaporative Concentrator
Clear seal joint
Concentration of trace amounts of analyte in organic solvent
ASTM E 438