missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Low-Profile Reservoirs

Replacement glass reservoirs for Acurex™ Low-Profile Dispensers

Brand:  DWK Life Sciences 844086

 View more versions of this product

Product Code. 12457201

  • £89.70 / Each
Estimated Shipment: 07-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Replacement 1000mL Amber Glass Reservoir
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Reservoir
Glass
Acurex™ low profile dispenser
Amber
1000 mL
33.81 oz.
Product Suggestions

Product Suggestions

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote