Learn More
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Brand: DWK Life Sciences W228781
16.25 GBP valid until 2024-09-30
Use promo code "21675" to get your promotional price.
This item is not returnable.
View return policy
Description
For compact and organized sample storage
- Stores vials in clear polystyrene case
- Foam inserts and partitions provide protection
- With alphanumerical indexing card for identification
- Index cards and partitions also available separately
Specifications
Storage and Transportation of Sample Vials | |
60 | |
6 Case |
228778 Hinged Case | |
Partition |