Learn More
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Brand:  DWK Life Sciences 228780
 View more versions of this product
Product Code. 10362663
  • £159.00 / Pack of 6
Explore more special offers
Description
Description
For compact and organized sample storage
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hinged Lid
Plastic
Convenient Storage and Transportation of Sample Vials
40 (2 and 4 mL Vial)
40
6 Case
Clear
Rectangular
40 x 4mL vials
Foam Insert, Index Card, Partition, Polystyrene Case
Storage Box