missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ 125mL Erlenmeyer Flask
With threaded mouth
Brand: DWK Life Sciences 635926
This item is not returnable.
View return policy
Description
Specifications
125 mL | |
Borosilicate Glass | |
24/40 |