Learn More
DWK Life Sciences Wheaton™ Arsine Generator, Modified
Modified for Arsine analysis
Brand: DWK Life Sciences 180023
This item is not returnable.
View return policy
Description
- Modified version of catalog number 180020 for arsine analysis
- The freestanding system has a standard taper 24/40 Clear-Seal™ joint
- One piece scrubber/absorber unit
- Low extractable borosilicate glass
- Eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes
- Manufactured from WHEATON 33 low-extractable borosilicate glass
Compliance
Conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Arsine Generator | |
Borosilicate Glass | |
For arsine analysis using SDDC method |
24/40 | |
USP Type I and ASTM E438 Type I, Class A | |
125mL erlenmeyer flask with 24/40 joint, one-piece scrubber/absorber unit |