Learn More
DWK Life Sciences Wheaton™ Arsine Scrubber Tube With 24/40, 12/2 Joint
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
Brand: DWK Life Sciences 180022
This item is not returnable.
View return policy
Description
- Scrubber tube
- Standard taper 24/40
- Clear-Seal joint and a spherical 12/2 ball joint held together by a size 12 stainless steel ball joint clamp
- Manufactured from WHEATON 33 low extractable borosilicate glass
Compliance
Meets ASTM / USP and EPA specifications; Borosilicate glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
| Arsine Scrubber Tube | |
| Borosilicate Glass | |
| For arsine analysis using SDDC method | |
| Size 12 stainless steel ball joint clamp |
| 24/40, 12/2 | |
| USP Type I and ASTM E438 Type I, Class A | |
| For arsine generator |
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Arsine Scrubber Tube With 24/40, 12/2 Joint > Scrubber Tube
Spot an opportunity for improvement?Share a Content Correction