missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Large Safety-Coated Solution and Serum Bottles

Product Code. 12482571 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
18,927 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Plastisol coated for safety.

  • Material: Borosilicate Glass
  • Not autoclavable
  • Shape: Round
  • Takes rubber stopper No. 8
  • Light resistant
TRUSTED_SUSTAINABILITY

Specifications

Capacity 640 oz., 18927 mL
Closure Material Rubber
Diameter (Metric) 294 mm
Height (English) 19.76 in.
Height (Metric) 502 mm
Product Type Serum Bottle
Capacity (Metric) 18,927 mL
Capacity (English) 640 oz.
Color Clear
Diameter (English) 11.57 in.
Material Borosilicate Glass
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.