Learn More
DWK Life Sciences Wheaton™ Arsine Generators
Assembled apparatus is freestanding and easy to use
Brand: DWK Life Sciences 180030
256.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Used for arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method.
- Manufactured from WHEATON 33 low-extractable borosilicate glass
The design of catalog No. 02912555 eliminates the ball and socket joint connecting the scrubber and absorber components
Compliance
Conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Arsine Generator | |
Borosilicate Glass | |
USP Type I and ASTM E438 Type I, Class A | |
For arsine analysis using SDDC method |
Product Suggestions
Customers who viewed this item also viewed