Learn More
DWK Life Sciences Wheaton™ Scrubber Absorber Tube
For arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method
Brand:  DWK Life Sciences 180034
71.90 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12445482
  • £99.20 / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Design eliminates the ball and socket joint connecting the scrubber and absorber components
  • Assembled apparatus is freestanding and easy to use
  • Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Applications:
For arsenic analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Scrubber/Absorber Tube
For arsenic analysis using silver diethyldithiocarbamate method
Borosilicate Glass
1/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Scrubber Absorber Tube > Scrubber/Absorber Tube