missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Serum Bottles and Vials

Product Code. 10021362
Click to view available options
Capacity:
0.1 oz., 2 mL
0.17 oz., 5 mL
0.17 oz., 5mL
0.34 oz., 10 mL
0.7 oz., 20 mL
1.01 oz., 30 mL
1.7 oz., 50 mL
2.03 oz., 60 mL
3.38 oz., 100 mL
4.23 oz., 125 mL
Color:
Amber
Clear
Height (English):
1.41 in.
1.85 in.
2.12 in.
2.28 in.
2.44 in.
2.48 in.
2.87 in.
3.58 in.
3.74 in.
4.21 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

These bottles have exceptional wall strength and superior heat and chemical tolerances

Ideal for long- and short-term sample storage and vaccine/injectable drug containers.

  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage

Compliance

Meet requirements for Class A of the United States Pharmacopeia for Type I glass.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 3.38 oz., 100 mL
Capacity (Metric) 100 mL
Capacity (English) 3.38 oz.
Color Clear
Diameter (English) 2.04 in.
Material Borosilicate Glass
Height (Metric) 95 mm
Height (English) 3.74 in.
Diameter (Metric) 52 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'