missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Serum Bottles and Vials
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
30 mL
50 mL
60 mL
100 mL
125 mL
Color:
Amber
Clear
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
- Clear borosilicate glass conforms to USP Type 1 requirements
- Amber borosilicate glass conforms to USP Type 1 requirements
- Shrink wrapped partitioned modules reduce breakage
Compliance
Meet requirements for Class A of the United States Pharmacopeia for Type I glass.

Specifications
Specifications
Capacity | 4.23 oz., 125 mL |
Capacity (Metric) | 125 mL |
Capacity (English) | 4.23 oz. |
Color | Clear |
Diameter (English) | 2.12 in. |
Material | Borosilicate Glass |
Height (Metric) | 107 mm |
Height (English) | 4.21 in. |
Diameter (Metric) | 54 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Documents
Scientific Resources
Catalogs
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ Serum Bottles and Vials >
Spot an opportunity for improvement?Share a Content Correction