missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Serum Vials
Available as clear and amber vials
£81.45 - £303.00
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | For lyophilization, packaging, sampling and freeze drying applications |
Autoclavable | Autoclavable |
Closure Type | Crimp Cap |
Certifications/Compliance | ASTM E438, USP Type I, Type I, Class A |
Product Code | Brand | Color | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | |||
10472752
|
DWK Life Sciences
223683 |
Clear | 15 mm | 2 mL | |||||
10747944
|
DWK Life Sciences
223684 |
Clear | 17 mm | 3 mL | |||||
10071332
|
DWK Life Sciences
223685 |
Clear | 22 mm | 5 mL |
£303.00
Pack of 144 |
In Stock
Log in to see stock. |
|||
10101952
|
DWK Life Sciences
223687 |
Clear | 30.5 mm | 20 mL |
£144.00
Pack of 120 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||
10757944
|
DWK Life Sciences
223693 |
Amber | 15 mm | 2 mL |
£102.00
Pack of 144 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
10623543
|
DWK Life Sciences
223695 |
Amber | 22 mm | 5 mL | |||||
10453522
|
DWK Life Sciences
223696 |
Amber | 24 mm | 10 mL |
£148.00
Pack of 144 |
In Stock
Log in to see stock. |
|||
10532313
|
DWK Life Sciences
223686 |
Clear | 24 mm | 10.0 mL | |||||
Description
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
- Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
- Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
- Withstand extreme temperature variations
- Necks finished for aluminum crimp seals
- Blowback feature of neck and specially designed bottom radius for lyophilization applications
- Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Compliance
Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards
Specifications
Borosilicate Glass | |
Autoclavable | |
ASTM E438, USP Type I, Type I, Class A |
For lyophilization, packaging, sampling and freeze drying applications | |
Crimp Cap |