missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Serum Vial
Serum vials are converted from Type I borosilicate glass tubing.
Brand: DWK Life Sciences™ WUBB3MLC-T-EP
This item is not returnable.
View return policy
Description
- Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
Specifications
0.28 in. | |
7 mm | |
13 mm | |
No | |
3 mL | |
0.1 oz. | |
Crimp Cap | |
169 Vials/Pk. | |
Serum Vial |
0.51 in. | |
Clear | |
Borosilicate Glass | |
Non-sterile | |
1.48 in. | |
Autoclavable | |
37.7 mm | |
USP Type I | |
No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences™ Wheaton™ Serum Vial > Serum Vials; Color: Clear; Volume (Metric): 3 mL; Quantity: 169 Vials/Pk.
Spot an opportunity for improvement?Share a Content Correction