missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Serum Vial
Serum vials are converted from Type I borosilicate glass tubing.
£108.62 - £245.41
Specifications
Material | Borosilicate Glass |
---|---|
Pre-slit | No |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Type | Serum Vial |
Product Code | Brand | Color | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
18215435
|
DWK Life Sciences™
WUBB2MLA-T-EP |
Amber | 2 mL | 225 Vials/Pk. |
£245.41
Pack of 225 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18205435
|
DWK Life Sciences™
WUBB2MLC-T-EP |
Clear | 2 mL | 225 Vials/Pk. |
£188.27
Pack of 225 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18235435
|
DWK Life Sciences™
WUBB3MLC-T-EP |
Clear | 3 mL | 169 Vials/Pk. |
£145.12
Pack of 169 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18265435
|
DWK Life Sciences™
WUBB3MLA-T-EP |
Amber | 3 mL | 169 Vials/Pk. |
£177.75
Pack of 169 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18255435
|
DWK Life Sciences™
WUBB5MLC-T-EP |
Clear | 5 mL | 100 Vials/Pk. |
£108.62
Pack of 100 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18245435
|
DWK Life Sciences™
WUBB5MLA-T-EP |
Amber | 5 mL | 100 Vials/Pk. |
£125.85
Pack of 100 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18275435
|
DWK Life Sciences™
WUBB10MLC-T-EP |
Clear | 10 mL | 150 Vials/Pk. |
£193.77
Pack of 150 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18225435
|
DWK Life Sciences™
WUBB10MLA-T-EP |
Amber | 10 mL | 150 Vials/Pk. |
£234.85
Pack of 150 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
Specifications
Borosilicate Glass | |
Non-sterile | |
Serum Vial |
No | |
Autoclavable | |
No |