missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Open-Top Seals
Open-top seal exposes stopper for sample retrieval with a syringe
Brand: DWK Life Sciences 224176-06
This item is not returnable.
View return policy
Description
- Unlined
- Closure Type: Seal

Specifications
Red | |
1000/Cs. | |
11 mm |
Aluminum | |
Open Top Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ Unlined Aluminum Open-Top Seals > 11mm (0.43in.); Red
Spot an opportunity for improvement?Share a Content Correction