missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ V Vial™ with Open-Top Screw Cap: Clear

Conical interior bottoms are designed to provide total downward drainage and maximum retrieval of contents by syringe

Brand:  DWK Life Sciences W986283NG

 View more versions of this product

Product Code. 11546305


Includes: Silicone-lined, black phenolic screw caps faced with PTFE.
This item is not returnable. View return policy

Description

Description

Manufactured from Wheaton-33™ low extractable borosilicate glass.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTFE/Silicone
13 mm
0.3 mL
No
Screw
Phenolic
35 mm
Clear
Borosilicate Glass (Low Extractable)
Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Conical V
0.1 dr.
Screw Cap (Open-top)
ASTM E438, USP Type I, Type I, Class A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers