missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners

Product Code. 11728909
Click to view available options
Neck Finish:
13-425
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
Material:
Glass
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

  • Liner provides tight seal
  • Liner Material: Foam, Polyethylene
  • Polypropylene
  • Screw Cap
  • White

Use these caps with glass bottles or vials

Order Info

Each pack contains 72

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Glass
No. per Case 72 Cs.
Neck Finish 53-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Closure Liner Polyethylene Foam
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'