missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
£57.60 - £288.00
Specifications
Color | White |
---|---|
Material | Polypropylene |
Certifications/Compliance | ISO 9001 |
For Use With (Equipment) | Glass bottle or vial |
Product Code | Brand | Neck Finish | Closure Liner | Material | No. per Case | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Neck Finish | Closure Liner | Material | No. per Case | Price | Quantity & Availability | ||
11784219
|
DWK Life Sciences
239229 |
20-400 | PTFE/Polyethylene | Polypropylene | 144 Cs. |
£57.60
Pack of 144 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11714229
|
DWK Life Sciences
239235 |
28-400 | PTFE/Polyethylene | Polypropylene | 144 Cs. |
£102.00
Pack of 144 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11708819
|
DWK Life Sciences
239243 |
63-400 | PTFE/Polyethylene | Polypropylene | 48 Cs. |
£146.00
Pack of 48 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11798809
|
DWK Life Sciences
239241 |
53-400 | PTFE/Polyethylene | Polypropylene | 72 Cs. |
£154.00
Pack of 72 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11704229
|
DWK Life Sciences
239233 |
24-400 | PTFE/Polyethylene | Polypropylene | 144 Cs. |
£76.20
Pack of 144 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
11778809
|
DWK Life Sciences
239238 |
43-400 | PTFE/Polyethylene | Polypropylene | 72 Cs. |
£104.00
Pack of 72 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11788809
|
DWK Life Sciences
239239 |
45-400 | Polyethylene Foam | Polypropylene | 72 Cs. |
£113.00
Pack of 72 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11724229
|
DWK Life Sciences
239236 |
33-400 | PTFE/Polyethylene | Polypropylene | 144 Cs. |
£120.00
Pack of 144 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11794219
|
DWK Life Sciences
239231 |
22-400 | Polyethylene Foam | Polypropylene | 144 Cs. |
£84.55
Pack of 144 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11734229
|
DWK Life Sciences
239240 |
48-400 | PTFE/Polyethylene | Polypropylene | 72 Cs. |
£135.00
Pack of 72 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
11744229
|
DWK Life Sciences
239242 |
58-400 | PTFE/Polyethylene | Polypropylene | 72 Cs. |
£172.00
Pack of 72 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11768809
|
DWK Life Sciences
239237 |
38-400 | PTFE/Polyethylene | Polypropylene | 72 Cs. |
£75.30
Pack of 72 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
10113052
|
DWK Life Sciences
239246 |
89-400 | PTFE/Polyethylene | Polypropylene | 48 Cs. |
£288.00
Pack of 48 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
11754229
|
DWK Life Sciences
239244 |
70-400 | PTFE/Polyethylene | Polypropylene | 48 Cs. |
£181.00
Pack of 48 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
Description
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
- PTFE faced foamed polyethylene liner offers chemical resistance
- Polyethylene foam provides compressibility and sealing properties
- Good for high purity chemicals, strong acids, and solvents
- For usewith glass bottles or vials
- Polypropylene
- Screw cap
- White
Specifications
White | |
Polypropylene | |
ISO 9001 | |
Glass bottle or vial |