missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ White 96-Well Immuno Plates
Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
£174.00 - £682.00
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Color | White |
Product Type | Microplate |
No. of Wells | 96 |
Product Code | Brand | Surface Treatment | Well Shape | Volume (Metric) Working | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Well Shape | Volume (Metric) Working | Price | Quantity & Availability | |||
10396181
|
Thermo Scientific™
436110 |
MaxiSorp | Round, Flat Bottom (F-well) | 350 μL |
£682.00
Pack of 80 |
In Stock
Log in to see stock. |
|||
10020581
|
Thermo Scientific™
436111 |
PolySorp | Round, Flat Bottom (F-well) | 350 μL |
£288.00
Pack of 80 |
In Stock
Log in to see stock. |
|||
11982607
|
Thermo Scientific™
7571 |
Microlite 1+ | Round, Flat Bottom (F-well) | 280 μL |
£265.00
Pack of 50 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10190725
|
Thermo Scientific™
7572 |
Microlite 2+ | Round, Flat Bottom (F-well) | 280 μL |
£213.00
Pack of 50 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||
10415985
|
Thermo Scientific™
7705 |
Microfluor 1 | Round, Flat Bottom (F-well) | 280 μL |
£174.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11952984
|
Thermo Scientific™
7905 |
Microfluor 2 | Round, Flat Bottom (F-well) | 280 μL |
£214.00
Pack of 50 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||
10537205
|
Thermo Scientific™
9502887 |
Universal Binding | Round, Flat Bottom (F-well) | 330 μL |
£357.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
- Plate has SBS footprint — fits standard equipment
- Available with flat- or round-bottom wells
- Choice of three surface types for optimal binding
- Certified binding homogeneity/reproducibility
Choice of Surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
- Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
- Gives maximum reflection
- Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
Specifications
Standard | |
White | |
96 |
8 x 12 | |
Microplate |