missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ White and Black 384-Well Immuno Plates
Conduct quantitative and qualitative immunoassays with these white and black 384-well immuno plates.
£428.00 - £692.00
Specifications
Binding Property | Hydrophilic |
---|---|
Format | Standard |
Lid | Without Lid |
Array | 16 x 24 |
Product Type | Microplate |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10385991
|
Thermo Scientific™
460372 |
White |
£428.00
Pack of 30 |
In Stock
Log in to see stock. |
|||||
10395991
|
Thermo Scientific™
460518 |
Black |
£692.00
Pack of 30 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Description
Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates. Ideal for ELISA and binding assays, these plates are compatible with common instruments. They offer lot-to-lot reliability and well-to-well reproducibility for optimal results.
Features:
- Plate has SBS footprint, fits standard equipment and is available with flat-bottom wells
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
- Plate is offered as black or white for optimal choice of application type
- Certified binding offers homogeneity/reproducibility.
White:
- Maximum reflection/minimum autofluorescence and autoluminescence
Black:
- Minimizes crosstalk, back-scattered light and background in fluorescence reading
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering information:
Availability may vary by country
Specifications
Hydrophilic | |
Without Lid | |
Microplate | |
384 | |
Non-sterile | |
Square, Flat Bottom (F-well) | |
10 | |
Antibodies (Glycoproteins) |
Standard | |
16 x 24 | |
Polystyrene | |
100 μL | |
MaxiSorp | |
30 | |
None | |
F384 |