Learn More
3M™ White/Blue Combined SMS Laminate Protective Coveralls
Product Code. 12952325
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Protective coveralls are manufactured from a high quality laminated material with a breathable back panel to improve air ventilation and reduce heat build-up.
  • Breathable back panel to help improve air ventilation and reduce heat build-up.
  • Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
  • Elastic waist and ankles for convenience and freedom of movement.
  • Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Color Blue, White
Disposable Single-use
Waist Style Elastic
Cuff Style Knit
Neck Style Hooded
Sterility Non-sterile
Protection Class Category III
Model 4535W