missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Cotton Wool, Absorbent

Test absorbancy by placing a teaspoon of water in the hollow of a cotton. Azpack™ Cotton Wool, Absorbant is BP quality.

Brand:  Azpack™ 12356477

39.64 GBP valid until 2024-12-31
Use promo code "21908" to get your promotional price.


Product Code. 12356477

  • £111.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 100% Bleached cotton combers
  • BP quality
  • 1 roll is approximately 3.5m x 325mm (L x W)
  • 500g per roll
Specifications

Specifications

Absorbent Cotton
BP Quality
500 g
Bleached Cotton
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.