Learn More
Buerkle™ White Disposable Forceps
GREENER_CHOICEBIOPHARMA
Product Code. 16525210 Shop All Bürkle Products
Click to view available options
Tip Style:
Angled, Pointed
Broad
Pointed
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 16525210
Brand: Buerkle™ 53780042
Only null left
Add to basket
Add to basket
Disposable forceps are ideal for grasping, lifting, and moving small objects like fine tissue or solid samples.
  • The fluted handles ensure optimum and secure handling as well as precise work
  • The light plastic forceps for single use allow for fatigue-free work
  • The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
  • The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
  • Sterile forceps are individually wrapped
  • For single use
  • Cleanroom manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
  • Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
  • Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
  • Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
TRUSTED_SUSTAINABILITY
Specifications
Tip Style Broad
No. per Pack 100 (large pack)
Length (English) 5.12 in.
Material Polystyrene
Length (Metric) 130 mm
Product Type Forceps
Sterility Non-sterile
Style Standard