missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White Disposable Visitor Lab Coat, Series 4400
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Breathable, lightweight material.
- Designed for light duty applications.
- Offers a barrier against non-hazardous particles and liquid splashes.
- Zip fastener.
- Anti-static treated.
- Elastic cuffs

Specifications
Specifications
Product Type | General Purpose Lab Coat |
Size | X-Large |
Gender | Unisex |
Material | Polypropylene |
Color | White |
Closure Type | Zipper |
Cuff Style | Elastic |
Disposable | Single-use |
Seam Type | Serged |
Size (English) Chest | 38 to 40 in. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ White Disposable Visitor Lab Coat, Series 4400 >
Spot an opportunity for improvement?Share a Content Correction