missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Visitor Lab Coat, Series 4400

Product Code. 12931335
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Offers a barrier against non-hazardous particles and liquid splashes. The Series 4400 Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.

  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Elastic cuffs
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Polypropylene
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Seam Type Serged
Size (English) Chest 28 to 32 in.
Size (Metric) Chest 71 to 81 cm
Certifications/Compliance (Europe) CE Category 1
For Use With (Application) Non-hazardous liquids and dust
Includes Internal pocket on the wearers left hand side (measuring 13 H x 12cm W)
Packaging Type Individually Packaged
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'