missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Visitor Lab Coat, Series 4400

Product Code. 12951335
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large

Offers a barrier against non-hazardous particles and liquid splashes. The Series 4400 Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.

  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Elastic cuffs
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Resistance Non-hazardous Liquids, Dust
Seam Type Serged
Size (English) Chest 32 to 36 in.
Size (Metric) Chest 81 to 91 cm
Certifications/Compliance (Europe) CE Category 1
For Use With (Application) Non-hazardous liquids and dust
Includes Internal pocket on the wearers left hand side (measuring 13 H x 12cm W)
Packaging Type Individually Packaged
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.