missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Visitor Lab Coat, Series 4400

Product Code. 15351495
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Offers a barrier against non-hazardous particles and liquid splashes. The Series 4400 Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.

  • Breathable, lightweight material
  • Designed for light duty applications
  • Offers a barrier against non-hazardous particles and liquid splashes
  • Zip fastener
  • Anti-static treated
  • Elastic cuffs
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lab Coat
Size 3X-Large
Material Microporous laminate
Color White
Cuff Style Elastic
Disposable Yes
Model Series 4400
For Use With (Application) Light duty applications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.