Learn More
PSF Progressive Safety™ White Hygiene Lace-Up Safety Work Shoes
An S2 SRC rated safety shoe, ideal for pharmaceuticals, food processing and catering, medical and laboratories, butcheries and fisheries.
Brand: PSF Progressive Safety™ W222-10
This item is not returnable.
View return policy
Description
- All safety footwear is designed with both comfort and durability in mind
- Conforms to EN ISO 20345:2011 for 200 joules protection
- Slip resistant sole
- Water resistant microfiber upper
- Machine washable
- Antibacterial lining
- Antistatic sole
Specifications
EN ISO 20345:2011 | |
Lace-up | |
Microfiber | |
Slip-resistant, antistatic |
White | |
Pharmaceutics, food industry, medical, laboratories, fisheries, butcheries | |
10 | |
Water, oils, fats, food waste |