missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ White Plastic General Purpose Trays
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
£56.30 - £121.00
Specifications
Color | White |
---|---|
Material | Plastic |
Type | Tray |
For Use With | Safe containment of spillages, lab bench protection |
Includes | Liner |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12347237
|
Fisherbrand™
|
460 x 260mm |
£56.30
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
12357237
|
Fisherbrand™
|
570 x 340mm |
£60.50
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
12367237
|
Fisherbrand™
|
570 x 540mm |
£85.97
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
12377237
|
Fisherbrand™
|
680 x 540mm |
£101.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
12387237
|
Fisherbrand™
|
700 x 460mm |
£104.23
Each |
In Stock
Log in to see stock. |
|||||
12397237
|
Fisherbrand™
|
1130 x 540mm |
£121.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- Available in general purpose, biohazard and radiation safety formats
- Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifications
White | |
Tray | |
Liner | |
Rectangular |
Plastic | |
Safe containment of spillages, lab bench protection | |
1 Each |