missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Polypropylene PE Foam/PTFE Cap

Product Code. 15513271
Click to view available options
Quantity:
100/Pk.
144EA/Cs.
288/PK.
500EA/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene PE Foam/PTFE Cap have a PTFE facing, backed by F217 liners.

Polypropylene (PP) caps with F217 & PTFE liners are an economical alternative to Thermoset & Phenolic PTFE lined caps. They have a PTFE facing, backed by F217 liners. The F217 liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE). PTFE/F217 liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference.

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference

Specifications

Color White
Description Screw cap
Material Polypropylene
No. per Case 100/Pk.
Packaging Packed in 5 bags of 100
Quantity 100/Pk.
Sterility Sterile
Closure Color White
Liner Material PE Foam/PTFE
Neck Finish 28-400
Certifications/Compliance ISO 9001
Closure Included Yes
Shape Round
Product Type PE Foam/PTFE Lined Cap
For Use With (Equipment) Bottle
Closure Liner PTFE/Polyethylene
Closure Material Polypropylene
Closure Size 28-400 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'