missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ White Polypropylene Caps
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Quantity:
100
144
288
370
500
1300
2100
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Color | White |
Quantity | 500 |
Closure Color | White |
Neck Finish | 15-425 |
Closure Included | No |
Product Type | Lined Cap |
Closure Liner | PTFE/Polyethylene Foam |
Closure Material | Polypropylene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'