missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White Disposable Visitor Lab Coat, Series 4400
Click to view available options
Lab Coat, Series 4400; Size: 2X-Large; Color: White
Description
- Breathable, lightweight material.
- Designed for light duty applications.
- Offers a barrier against non-hazardous particles and liquid splashes.
- Zip fastener.
- Anti-static treated.
- Knitted cuffs.
Specifications
Product Type | General Purpose Lab Coat |
Size | 2X-Large |
Gender | Unisex |
Material | Polypropylene |
Color | White |
Closure Type | Zipper |
Cuff Style | Elastic |
Disposable | Single-use |
Resistance | Non-hazardous Liquids, Dust |
Seam Type | Serged |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'