missing translation for 'onlineSavingsMsg'
Learn More
Learn More
White Swing Bin Liner 33 x 58 x 76cm
Brand: N/A 070004
This item is not returnable.
View return policy
Description
• White• Size 330 x 584 x 736mm