missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ White Type 5/6 Air Disposable Chemical Protection Coveralls
242.76 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Exceptional wearer comfort guaranteed by breathable and lightweight SMS material
- Particle-tight and spray-tight to a limited extent
- Ideal for warm working environments and extended periods of wear
- Free of silicone and paint wetting impairment substances
- Material: Polypropylene SMS
- Middle finger loops prevent sleeves sliding up the arm
- Optimal protection in dusty environments
- Self-adhesive zipper flap for optimal protection
- Anti-static properties
- Excellent moisture management
- Optimal fit with elasticated waistband
- Three-part hood for a good fit and unrestricted vision
- Secure and convenient closures thanks to elasticated bands on hood, arms and legs
Specifications
Product Type | Cleanroom Coveralls |
Size | Small |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Certifications/Compliance | ISO 13982-1 - type 5, EN 13034 - type 6, EN 1149-5, EN 1073-2 |
For Use With (Application) | Controlled Environments |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'