missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ White Urea Screw Caps

Product Code. p-8002161 Shop All Wheaton Science Products Products
Click to view available options
Closure Size:
15.5 mm
22 mm
Unit Size:
Pack of 1000

Product Code. 14813592

Brand: Wheaton™ 300190

Only null left
Add to basket
Add to basket

Suitable for most applications that call for screw neck bottles. Wheaton™ White Urea Screw Caps are EPE lined.

  • Multiple thread size options
  • Excellent closing option for screw neck bottles and vials
TRUSTED_SUSTAINABILITY

Specifications

Closure Size 15.5 mm
Liner Material EPE
Description Cap
Material Urea
Color White
Quantity 1000 Pack
For Use With (Equipment) Screw Thread Diagnostic Vials
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.