Learn More
Azlon™ Wide Mouth Vented Wash Bottles Product Code.: 15347047
In Stock
to see stock.
Add to basket
Azlon™ Wide Mouth Vented Wash Bottles
Product Code. 15347047
500 mL, Green
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Color:
Blue
Green
Orange
Red
White
Yellow
Product Code. 15347047
Brand: Azlon WGW239PVENT
In Stock
to see stock.
Edge
Add to basket
Add to basket
Manufactured from LPDE for strength and flexibilty. Azlon™ Wide Mouth Vented Wash Bottles feature color-coded lids for easy identification.
Manufactured from LPDE for strength and flexibilty. Azlon™ Wide Mouth Vented Wash Bottles feature color coded lids for easy identification.
  • Square shoulder wash bottles
  • Wide neck for easy spill-free filing; patented DripLok™  vapor venting valve prevents solvent drips
  • Color-coded one-piece polypropylene closures maintain shape in use
  • Precision jet-tip aperture for fine control
  • Translucent bottles without print
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Material LDPE
Closure Color Green
Mouth Wide
Closure Material Polypropylene
Closure Type Screw Cap with Curved Aperture
Product Content Correction
Product Title
Azlon™ Wide Mouth Vented Wash Bottles >