Learn More
BRAND™ Wide Neck Glass Erlenmeyer Flasks
With beaded rim and graduations
£96.25 - £219.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10089860
|
BRAND™
92836 |
250 mL |
£109.00
Pack of 10 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10141681
|
BRAND™
92817 |
50 mL |
£99.55
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10151871
|
BRAND™
92844 |
500 mL |
£140.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10293391
|
BRAND™
92824 |
100 mL |
£96.25
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10419912
|
BRAND™
92839 |
300 mL |
£127.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10694352
|
BRAND™
92854 |
1000 mL |
£219.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Manufactured from Boro 3.3 glass
- Wide neck for ease of filling
- Acc. to DIN EN ISO 24450
- Provided in packs of 10
Specifications
Borosilicate Glass (3.3) | |
10 |