missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Wide mouth, Short form DIN thread Product Code.: 12891004
Add to basket

DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Wide mouth, Short form DIN thread

Product Code. 12891004 Shop All DWK Life Sciences Products
100 mL
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
This item is not returnable. View return policy

Product Code. 12891004

Brand: DWK Life Sciences 238202405

Add to basket
Add to basket
This item is not returnable. View return policy

Square clear soda lime glass bottles, wide mouth.

Square clear soda lime (Type 3) glass bottles, wide mouth and short form DIN thread.

  • Square shape for efficient packing
  • Wide bottle neck allows easy access
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 mL
For Use With (Application) Storage of samples and reagents; Sample collection; Transport and shipping
Material Soda Lime Glass
Quantity 10/Pk.
Thread Size (Metric) GL 32
Autoclavable Not Autoclavable
Diameter (Metric) 49 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Wide mouth, Short form DIN thread >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.