Learn More
Quadralene Wide Spectrum Biocide Biocleanse Wipes
Ready-to-use wipes impregnated with a wide spectrum biocidal agent that is non-irritating and is effective against organisms including Listeria, Candida, C. Diff and Influenza. Also removes fat, grease, blood, protein and other contaminants.
Brand: Quadralene D/0007/90
Code : 24
Additional Details : CAS Number : 5064-31-3 Weight : 0.80100kg
Description
- Powerful bactericidal and virucidal properties
- Biocleanse is free of sodium hypochlorite, phosphates and enzymes
- Removes organic and inorganic compounds whilst simultaneously disinfecting the treated surface
- Safe to use on non-ferrous metals, ceramics, glassware and plastics
- Each wipe is 200 x 220mm, impregnated with 5% solution of Biocleanse concentrate
Specifications
Cleaning Wipe | |
Disinfecting and cleaning | |
5% of Biocleanse Concentrate |
200 x 220 mm | |
200/Pk. |