missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Wildco™ Kemmerer Water Sampler

Product Code. p-7116309 Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
400 mL
1200 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11774499

Brand: Cole-Parmer™ 0548500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sample at specific depths between 3 and 600 feet.

  • WildcoKemmerer Water Sampler
  • The messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle
  • All models include a plastic carrying case; order messenger and line separately.
  • The 304 SS models have urethane end seals (do not use when mercury concentrations exceed 1 ppm).

Specifications

Product Type Kemmerer Water Sampler
Material Stainless Steel
Description 3 to 600 ft depth
Capacity (Metric) 400 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.