missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ William's E Medium, GlutaMAX™ Supplement
GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
£42.82 - £325.00
Specifications
Cell Type | Mammalian Cells |
---|---|
Culture Type | Mammalian Cell Culture |
Form | Liquid |
Product Type | William's E Medium |
Without Additives | No HEPES |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11514466
|
Gibco™
32551087 |
10 x 500 mL |
£325.00
Pack of 10 |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10043282
|
Gibco™
32551020 |
500 mL |
£42.82
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells
- Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells
- Medium contains unique ingredients including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate
- GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
Modified as follows
- With: GlutaMAX™, phenol red
- Without: HEPES
Using Willam's E Medium
- Medium contains no proteins or growth factors therefore it requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS)
- William's E Medium uses a sodium bicarbonate buffer system (2.2 g/L)
- It requires a 5-10% CO2 environment to maintain physiological pH
Specifications
Mammalian Cells | |
Liquid | |
No HEPES |
Mammalian Cell Culture | |
William's E Medium |