missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ 5mm O.D. Thin Walled High-Throughput Economy NMR Tubes

Designed for routine use in all NMR spectrometers, Wilmad Economy NMR Tubes fit tightly in any spinner turbine and have minimal NMR background.

£12.57 - £15.15

Specifications

Material Borosilicate Glass
Diameter (Metric) Outer 4.947±0.019 mm
Field Strength 100MHz
For Use With (Application) Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only.
Includes Tube and disposable cap
View More Specs

Alert:

Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.


Products 3
Product Code Brand Model Length (English) Price Quantity & Availability  
Product Code Brand Model Length (English) Price Quantity & Availability  
15318668
View accessory products View Documents
Bel-Art™
66000-0084
Wilmad™ WG-5MM-ECONOMY-8-5 8 in.
£15.15
Pack of 5
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
 
15398658
View accessory products View Documents
Bel-Art™
66000-0083
Wilmad™ WG-5MM-ECONOMY-7-5 7 in.
£14.65
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
15338668
View Documents
Bel-Art™
66000-0085
Wilmad™ WG-5MM-ECONOMY-9-5 9 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • High quality 5mm O.D. economy sample tube
  • Best O.D. tolerance in the industry and a 30% thicker wall than any competitors' product
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
  • Fit tightly in any spinner turbine
  • Minimal NMR background
  • Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A
  • Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes
  • Manufactured in an ISO 9001:2008 USA facility
Specifications

Specifications

Borosilicate Glass
100MHz
Tube and disposable cap
5/Pk.
4.947±0.019 mm
Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only.
0.43 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.