Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Kel-F Positioning Rod Microcell Small Volume NMR Insert
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Kel-F Positioning Rod Microcell Small Volume NMR Insert is ideal for experiments that need to center small volume samples in the Rf coil such as magnetic susceptibility measurement.
Brand:  Bel-Art™ SP Scienceware™ 66000-0010
Product Code. 15328578
  • £48.50 / Each
Explore more special offers
Description
Description
  • Sample volume can be as little as 18 μl
  • 3 different inserts with various shapes (Spherical Bulb, Capillary Tube and Cylindrical Bulb)
  • Outer tube (Precision Thin Walled) must be ordered separately depending on magnetic field strength.
  • Not recommended for variable temperature experiments.
  • Use our PTFE Needles (#90630) with syringes to fill and clean sample.
Specifications
Specifications
529-B adapter, Vortex Plugs WG-805C and under
Positioning Rod for inserting small volume microcells
529-C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Only use with Wilmad-LabGlass Precision Thin-Walled Tubes