Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ O-Ring for Spinner
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ O-Ring for Spinner is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Brand: Bel-Art™ SP Scienceware™ 66000-0057
Code : 03
Additional Details : Weight : 0.12920kg
Description
- Low cost alternative to the full VT version for sample changers.
- Weight and susceptibility matched to originals.
- Can be mixed with originals in sample changer operation.
- VT operation of -150°C to 200°C
- Does not jam at top of upper barrel during insert operation.
- Tachometer strip now with foil protection for improved life span.
- Weight compatible with room temperature spinners.
- Very cost effective as compared to the originals.
Specifications
Agilent/Varian NMR Spectrometer | |
V-GFK-10 |