missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ PTFE/Silicone Rubber Septum for 4 to 5 mm Tubes
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ PTFE/Silicone Rubber Septum for 4 to 5 mm Tubes are commonly used in sample degasification.
Brand: Bel-Art™ SP Scienceware™ 66000-0052
Code : 03
Additional Details : Weight : 0.04310kg
Description
- Comes complete with open top screw cap and PTFE/Silicone septum
- Inert to most organic solvents and compounds but poor to strongly corrosive materials
- Remains reliable after multiple punctures
Specifications
Septum | |
4 mm and 5 mm Screw Cap NMR Tubes | |
36 Pieces |
PTFE, Silicone | |
Replacement Septum Only | |
TR-LS-01 |