Learn More
SP Wilmad-LabGlass™ High-Throughput NMR Tubes
Double the writing area allows for easy sample marking and identification.
Brand:  SP 66999-5751
 View more versions of this product
Product Code. 16695672
  • £154.00 / Pack of 50
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Average camber of 60 microns ensures spectrum clarity for small molecule (MW <250) samples up to 600 MHz.
  • 2x larger marking area eliminates cramped and unreadable sample names
  • Silkscreened writing area keeps sample names from easily wiping off
  • Designed for routine use in most low to mid field NMR spectrometers
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimensions
Note: For experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Specifications
Specifications
5±0.02 mm
7 in.
0.43 mm
50 Pack